Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUDP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416851.6 | XP_416851.3 | 3 - 4 | 593 | 98 | |
GenBank mRNA | BX932630.1 | - | 3 - 4 | 581 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |