Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277343.1 | NP_001264272.1 | 4 - 5 | 419 | 83 | |
GenBank mRNA | BX934192.1 | - | 3 - 4 | 393 | 83 | |
BX934456.2 | - | 4 - 5 | 405 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |