Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004940682.3 | XP_004940739.1 | 4 - 5 | 4505 | 97 | |
XM_004940683.3 | XP_004940740.1 | 3 - 4 | 4469 | 97 | ||
GenBank mRNA | BX935527.1 | - | 5 - 6 | 641 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |