Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277920.1 | NP_001264849.1 | 4 - 5 | 483 | 66 | |
GenBank mRNA | BX930994.2 | - | 4 - 5 | 483 | 66 | |
BX935748.1 | - | 4 - 5 | 485 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |