Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414755.5 | XP_414755.2 | 5 - 6 | 1232 | 74 | |
GenBank mRNA | BX934565.2 | - | 3 - 4 | 394 | 74 | |
BX936040.1 | - | 3 - 4 | 402 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |