Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTSSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291671.2 | XP_015147157.1 | 3 | 880 | 138 | |
XM_015291672.2 | XP_015147158.1 | 3 | 807 | 138 | ||
XM_015291673.2 | XP_015147159.1 | 3 | 839 | 138 | ||
XM_015291674.2 | XP_015147160.1 | 3 | 1013 | 138 | ||
GenBank mRNA | CR353390.1 | - | 2 | 548 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |