Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195594.1 | NP_001182523.1 | 3 - 4 | 455 | 90 | |
GenBank mRNA | BX931314.2 | - | 3 - 4 | 444 | 90 | |
BX935522.1 | - | 3 - 4 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |