Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277740.1 | NP_001264669.1 | 2 - 3 | 332 | 91 | |
XM_025154831.1 | XP_025010599.1 | 2 - 3 | 190 | 91 | ||
GenBank mRNA | AJ441614.1 | - | 2 - 3 | 165 | 91 | |
BX932212.2 | - | 2 - 3 | 231 | 91 | ||
CN232875.1 | - | 2 - 3 | 333 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |