Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199584.1 | NP_001186513.1 | 1 - 2 | 310 | 69 | |
XM_015286372.2 | XP_015141858.1 | 2 - 3 | 494 | 69 | ||
XM_015286373.2 | XP_015141859.1 | 5 - 6 | 2300 | 69 | ||
XM_025150668.1 | XP_025006436.1 | 2 - 3 | 499 | 69 | ||
XM_025150669.1 | XP_025006437.1 | 5 - 6 | 2300 | 69 | ||
GenBank mRNA | BX934209.2 | - | 1 - 2 | 310 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |