Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420705.6 | XP_420705.1 | 5 - 6 | 657 | 152 | |
GenBank mRNA | CR353754.1 | - | 4 - 5 | 455 | 152 | |
CR385517.1 | - | 5 - 6 | 395 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |