Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012593.2 | NP_001012611.2 | 23 - 24 | 2473 | 82 | |
XM_015288386.2 | XP_015143872.1 | 23 - 24 | 2549 | 82 | ||
XM_015288389.2 | XP_015143875.1 | 23 - 24 | 2549 | 82 | ||
XM_015288390.2 | XP_015143876.1 | 23 - 24 | 2543 | 82 | ||
XM_025151585.1 | XP_025007353.1 | 23 - 24 | 2549 | 82 | ||
XM_025151586.1 | XP_025007354.1 | 23 - 24 | 2546 | 82 | ||
XM_025151587.1 | XP_025007355.1 | 23 - 24 | 2543 | 82 | ||
XM_025151588.1 | XP_025007356.1 | 22 - 23 | 2492 | 82 | ||
XM_025151589.1 | XP_025007357.1 | 22 - 23 | 2375 | 82 | ||
GenBank mRNA | AJ851808.1 | - | 23 - 24 | 2475 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |