Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_418271.6 | XP_418271.3 | 1 | 1323 | 79 | |
GenBank mRNA | CR389334.1 | - | 1 | 252 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |