Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199532.1 | NP_001186461.1 | 3 - 4 | 249 | 129 | |
XM_015288099.1 | XP_015143585.1 | 3 - 4 | 227 | 129 | ||
XM_025151515.1 | XP_025007283.1 | 2 - 3 | 159 | 129 | ||
GenBank mRNA | BX932673.1 | - | 3 - 4 | 249 | 129 | |
CR387172.1 | - | 3 - 4 | 413 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |