Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199532.1 | NP_001186461.1 | 7 - 8 | 776 | 144 | |
XM_015288099.1 | XP_015143585.1 | 7 - 8 | 754 | 144 | ||
XM_025151515.1 | XP_025007283.1 | 6 - 7 | 686 | 144 | ||
GenBank mRNA | BU421613.1 | - | 2 - 3 | 312 | 144 | |
BX932673.1 | - | 7 - 8 | 776 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |