Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277593.1 | NP_001264522.1 | 2 - 3 | 247 | 61 | |
GenBank mRNA | BX933371.1 | - | 2 - 3 | 247 | 61 | |
CD727791.1 | - | 2 - 3 | 229 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |