Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277866.1 | NP_001264795.1 | 1 | 267 | 95 | |
GenBank mRNA | BU270193.1 | - | 1 | 277 | 95 | |
BX935000.1 | - | 1 | 249 | 95 | ||
CR386978.1 | - | 1 | 277 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |