Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199554.1 | NP_001186483.1 | 4 - 5 | 473 | 145 | |
XM_015281404.2 | XP_015136890.1 | 4 - 5 | 485 | 145 | ||
XM_025147802.1 | XP_025003570.1 | 3 - 4 | 513 | 145 | ||
XM_025147804.1 | XP_025003572.1 | 4 - 5 | 546 | 145 | ||
XM_025147808.1 | XP_025003576.1 | 4 - 5 | 432 | 145 | ||
XM_025147815.1 | XP_025003583.1 | 4 - 5 | 533 | 145 | ||
XM_025147817.1 | XP_025003585.1 | 4 - 5 | 439 | 145 | ||
XM_025147823.1 | XP_025003591.1 | 5 - 6 | 549 | 145 | ||
XM_025147830.1 | XP_025003598.1 | 4 - 5 | 569 | 145 | ||
GenBank mRNA | BU356502.1 | - | 4 - 5 | 446 | 145 | |
BU400702.1 | - | 2 - 3 | 220 | 145 | ||
BU409185.1 | - | 4 - 5 | 501 | 145 | ||
CR352492.1 | - | 4 - 5 | 476 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |