Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015290549.2 | XP_015146035.1 | 12 | 2538 | 62 | |
XM_422290.4 | XP_422290.2 | 12 | 2909 | 62 | ||
GenBank mRNA | CR352776.1 | - | 1 | 650 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |