Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252144.3 | NP_001239073.1 | 2 - 3 | 225 | 91 | |
GenBank mRNA | BX933738.1 | - | 2 - 3 | 225 | 91 | |
CR353787.1 | - | 3 - 4 | 345 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |