Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244905.1 | NP_001231834.1 | 446 | 98 | ||
GenBank mRNA | CR354169.1 | - | 187 | 98 | ||
CR523581.1 | - | 246 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus |