Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277630.1 | NP_001264559.1 | 8 - 9 | 938 | 70 | |
NM_001277631.1 | NP_001264560.1 | 7 - 8 | 818 | 70 | ||
XM_025148693.1 | XP_025004461.1 | 8 - 9 | 923 | 70 | ||
GenBank mRNA | BU224670.1 | - | 4 - 5 | 443 | 70 | |
BX929977.2 | - | 7 - 8 | 802 | 70 | ||
BX934152.2 | - | 8 - 9 | 939 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |