Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015286685.2 | XP_015142171.1 | 7 - 8 | 1048 | 157 | |
GenBank mRNA | BX934822.2 | - | 7 - 8 | 988 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |