Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277603.1 | NP_001264532.1 | 4 - 5 | 500 | 64 | |
GenBank mRNA | BX934878.2 | - | 4 - 5 | 394 | 64 | |
BX935074.2 | - | 4 - 5 | 500 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |