Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004948703.3 | XP_004948760.1 | 7 - 8 | 1188 | 74 | |
XM_418193.6 | XP_418193.3 | 7 - 8 | 1169 | 74 | ||
GenBank mRNA | CR352713.1 | - | 5 - 6 | 614 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |