Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302170.1 | NP_001289099.1 | 4 - 5 | 248 | 157 | |
NM_001302171.1 | NP_001289100.1 | 4 - 5 | 341 | 157 | ||
GenBank mRNA | CR354370.1 | - | 4 - 5 | 248 | 157 | |
CR387219.1 | - | 4 - 5 | 341 | 157 | ||
CR391062.1 | - | 4 - 5 | 369 | 157 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |