Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015287767.2 | XP_015143253.1 | 2 - 3 | 873 | 171 | |
XM_025150934.1 | XP_025006702.1 | 3 - 4 | 931 | 171 | ||
GenBank mRNA | BX933079.2 | - | 1 - 2 | 771 | 171 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |