Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942966.3 | XP_004943023.1 | 17 - 18 | 1782 | 122 | |
XM_422119.6 | XP_422119.4 | 16 - 17 | 1757 | 122 | ||
GenBank mRNA | BX950354.2 | - | 7 - 8 | 698 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |