Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143342.1 | XP_024999110.1 | 3 - 4 | 3103 | 70 | |
XM_025143343.1 | XP_024999111.1 | 3 - 4 | 3103 | 70 | ||
XM_025143344.1 | XP_024999112.1 | 3 - 4 | 3103 | 70 | ||
XM_025143345.1 | XP_024999113.1 | 4 - 5 | 2563 | 70 | ||
XM_025143346.1 | XP_024999114.1 | 3 - 4 | 5473 | 70 | ||
GenBank mRNA | AJ011002.1 | - | 2 - 3 | 2742 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |