Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLCD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277557.3 | NP_001264486.1 | 5 - 6 | 554 | 76 | |
XM_015290629.2 | XP_015146115.1 | 7 - 8 | 940 | 76 | ||
XM_025152828.1 | XP_025008596.1 | 6 - 7 | 721 | 76 | ||
XM_025152829.1 | XP_025008597.1 | 5 - 6 | 654 | 76 | ||
XM_025152830.1 | XP_025008598.1 | 5 - 6 | 829 | 76 | ||
XM_025152831.1 | XP_025008599.1 | 5 - 6 | 1255 | 76 | ||
GenBank mRNA | BX934812.1 | - | 5 - 6 | 554 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |