Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288444.2 | XP_015143930.1 | 9 - 10 | 844 | 71 | |
XM_421646.6 | XP_421646.1 | 9 - 10 | 750 | 71 | ||
GenBank mRNA | BX935521.1 | - | 10 - 11 | 752 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |