Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA8L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015288444.2 | XP_015143930.1 | 3 - 4 | 307 | 85 | |
XM_421646.6 | XP_421646.1 | 3 - 4 | 213 | 85 | ||
GenBank mRNA | BX935521.1 | - | 4 - 5 | 215 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |