Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM213A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193518.1 | NP_001180447.1 | 3 - 4 | 401 | 87 | |
NM_001193519.2 | NP_001180448.1 | 3 - 4 | 392 | 87 | ||
GenBank mRNA | AJ720950.1 | - | 3 - 4 | 393 | 87 | |
BU119844.1 | - | 3 - 4 | 297 | 87 | ||
BU489653.1 | - | 3 - 4 | 402 | 87 | ||
KP278239.1 | - | 2 - 3 | 271 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |