Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641978.4 | XP_003642026.1 | 1 - 2 | 92 | 98 | |
GenBank mRNA | CR406454.1 | - | 1 - 2 | 93 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |