Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199231.2 | NP_001186160.1 | 3 | 366 | 117 | |
XM_015277369.2 | XP_015132855.1 | 3 | 354 | 117 | ||
XM_015277370.2 | XP_015132856.1 | 3 | 334 | 117 | ||
GenBank mRNA | AJ453243.1 | - | 3 | 368 | 117 | |
AJ851491.1 | - | 3 | 342 | 117 | ||
BI394192.1 | - | 2 | 299 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |