Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005798.1 | NP_001005798.1 | 4 - 5 | 1135 | 64 | |
XM_015283067.2 | XP_015138553.2 | 4 - 5 | 1204 | 64 | ||
GenBank mRNA | AB105810.1 | - | 4 - 5 | 834 | 64 | |
AJ719936.1 | - | 4 - 5 | 1135 | 64 | ||
CR388496.1 | - | 4 - 5 | 755 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |