Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM206A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006383.1 | NP_001006383.1 | 3 - 4 | 265 | 142 | |
XM_015282375.2 | XP_015137861.1 | 3 - 4 | 436 | 142 | ||
XM_025147280.1 | XP_025003048.1 | 4 - 5 | 448 | 142 | ||
GenBank mRNA | AJ721017.1 | - | 3 - 4 | 265 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |