Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006481.1 | NP_001006481.1 | 4 - 5 | 615 | 125 | |
XM_015287388.2 | XP_015142874.1 | 4 - 5 | 125 | |||
GenBank mRNA | AJ719909.1 | - | 4 - 5 | 615 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |