Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204549.1 | NP_989880.1 | 10 - 11 | 1096 | 64 | |
XM_015291368.2 | XP_015146854.1 | 10 - 11 | 1118 | 64 | ||
GenBank mRNA | AF515726.1 | - | 10 - 11 | 1096 | 64 | |
AJ720280.1 | - | 10 - 11 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |