Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204554.1 | NP_989885.1 | 3 - 4 | 650 | 93 | |
XM_015292544.2 | XP_015148030.1 | 3 - 4 | 906 | 93 | ||
GenBank mRNA | AJ438290.1 | - | 3 - 4 | 650 | 93 | |
BX930581.2 | - | 3 - 4 | 488 | 93 | ||
BX931497.1 | - | 3 - 4 | 501 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |