Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAIL-LIKE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204591.2 | NP_989922.1 | 4 - 5 | 584 | 77 | |
XM_015278184.2 | XP_015133670.1 | 4 - 5 | 575 | 77 | ||
GenBank mRNA | AY057941.1 | - | 4 - 5 | 588 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |