Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204647.1 | NP_989978.1 | 3 - 4 | 3352 | 94 | |
XM_015275833.2 | XP_015131319.1 | 4 - 5 | 4808 | 94 | ||
XM_025147028.1 | XP_025002796.1 | 3 - 4 | 4708 | 94 | ||
GenBank mRNA | AF304358.1 | - | 3 - 4 | 3352 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |