Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204881.1 | NP_990212.1 | 1 - 2 | 591 | 124 | |
GenBank mRNA | AJ012219.1 | - | 1 - 2 | 591 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |