Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204881.1 | NP_990212.1 | 5 - 6 | 1207 | 90 | |
GenBank mRNA | AJ012219.1 | - | 5 - 6 | 1207 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |