Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204887.1 | NP_990218.1 | 3 - 4 | 669 | 82 | |
XM_015292954.2 | XP_015148440.1 | 3 - 4 | 1256 | 82 | ||
GenBank mRNA | AB006014.1 | - | 3 - 4 | 669 | 82 | |
KX897002.1 | - | 2 - 3 | 289 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |