Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205358.1 | NP_990689.2 | 1 - 2 | 140 | 142 | |
GenBank mRNA | L39880.1 | - | 1 - 2 | 140 | 142 | |
U41823.1 | - | 1 - 2 | 65 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |