Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205364.1 | NP_990695.1 | 1 - 2 | 265 | 66 | |
GenBank mRNA | U48860.1 | - | 1 - 2 | 265 | 66 | |
X83889.1 | - | 124 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |