Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205377.1 | NP_990708.1 | 5 - 6 | 399 | 81 | |
XM_015279338.2 | XP_015134824.1 | 5 - 6 | 1540 | 81 | ||
XM_015279339.2 | XP_015134825.1 | 5 - 6 | 516 | 81 | ||
GenBank mRNA | BX934267.1 | - | 5 - 6 | 519 | 81 | |
L27420.1 | - | 5 - 6 | 399 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |