Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205424.1 | NP_990755.1 | 3 - 4 | 302 | 127 | |
GenBank mRNA | L10920.1 | - | 1 - 2 | 62 | 127 | |
L12144.1 | - | 3 - 4 | 302 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |