Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205478.2 | NP_990809.2 | 6 - 7 | 807 | 101 | |
GenBank mRNA | CR388911.1 | - | 6 - 7 | 794 | 101 | |
M29449.1 | - | 6 - 7 | 807 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |