Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012570.1 | NP_001012588.1 | 6 - 7 | 629 | 128 | |
XM_015295391.2 | XP_015150877.1 | 5 - 6 | 534 | 128 | ||
GenBank mRNA | AJ851816.1 | - | 6 - 7 | 629 | 128 | |
BX929298.1 | - | 5 - 6 | 513 | 128 | ||
BX929381.1 | - | 5 - 6 | 526 | 128 | ||
BX933636.2 | - | 5 - 6 | 520 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |